2,3-Difluor-4-formylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2,3-Difluoro-4-formylphenyl)boronic acid
BB-8013-1G
COBBBB-8013-1GEA
0
EUR
COBBBB-8013-1G
2,3-Difluor-4-formylphenylboronsäure
2,3-Difluor-4-formylphenylboronsäure
Formel:
C₇H₅BF₂O₃ |
MDL:
MFCD04039907 CAS-Nummer: 480424-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...