3-Ethoxy-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8018-1G BB-8018-5G
COBBBB-8018-1GEA 37.5 EUR
COBBBB-8018-1G COBBBB-8018-5G
3-Ethoxy-2-fluorphenylboronsäure
3-Ethoxy-2-fluorphenylboronsäure
Formel: C₈H₁₀BFO₃
Molecular Weight: 183.97 g/mol
MDL: MFCD06798077
CAS-Nummer: 855230-61-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR