2,3,6-Trifluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8021-1G
COBBBB-8021-1GEA 25.5 EUR
COBBBB-8021-1G
2,3,6-Trifluorphenylboronsäure
2,3,6-Trifluorphenylboronsäure
Formel: C₆H₄BF₃O₂
Molecular Weight: 175,9 g/mol
MDL: MFCD01863167
CAS-Nummer: 247564-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR