2-Chlor-6-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8045-1G
COBBBB-8045-1GEA 0 EUR
COBBBB-8045-1G
2-Chlor-6-methoxyphenylboronsäure
2-Chlor-6-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
MDL: MFCD04039878
CAS-Nummer: 385370-80-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR