2-Benzyloxy-3-bromo-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Benzyloxy)-3-bromo-5-methylphenyl)boronic acid

BB-8048-1G
COBBBB-8048-1GEA 69 EUR
COBBBB-8048-1G
2-Benzyloxy-3-bromo-5-methylphenylboronsäure
2-Benzyloxy-3-bromo-5-methylphenylboronsäure
Formel: C₁₄H₁₄BBrO₃
Molecular Weight: 320,97 g/mol
MDL: MFCD06200723
CAS-Nummer: 870777-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR