2-Propoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8086-1G
COBBBB-8086-1GEA 98.3 EUR
COBBBB-8086-1G
2-Propoxyphenylboronsäure
2-Propoxyphenylboronsäure
Formel: C₉H₁₃BO₃
Molecular Weight: 180,0087 g/mol
MDL: MFCD03427191
CAS-Nummer: 134896-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR