2-Fluor-6-iodphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8090-1G
COBBBB-8090-1GEA 0 EUR
COBBBB-8090-1G
2-Fluor-6-iodphenylboronsäure
2-Fluor-6-iodphenylboronsäure
Formel: C₆H₅BFIO₂
MDL: MFCD06798080
CAS-Nummer: 870777-22-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR