4-[(1-Naphthyloxy)methyl]phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8124-1G
COBBBB-8124-1GEA 0 EUR
COBBBB-8124-1G
4-[(1-Naphthyloxy)methyl]phenylboronsäure
4-[(1-Naphthyloxy)methyl]phenylboronsäure
Formel: C₁₇H₁₅BO₃
MDL: MFCD08276787
CAS-Nummer: 871125-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR