4-(2'-Fluorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8130-1G
COBBBB-8130-1GEA 86.3 EUR
COBBBB-8130-1G
4-(2'-Fluorbenzyloxy)phenylboronsäure
4-(2'-Fluorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BFO₃
MDL: MFCD08705274
CAS-Nummer: 1072951-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR