2,6-Difluor-3-formylphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8152-1G
COBBBB-8152-1GEA 303 EUR
COBBBB-8152-1G
2,6-Difluor-3-formylphenylboronsäure
2,6-Difluor-3-formylphenylboronsäure
Formel: C₇H₅BF₂O₃
MDL: MFCD07369756
CAS-Nummer: 849062-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR