4-(2'-Methoxybenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8155-1G
COBBBB-8155-1GEA 93.7 EUR
COBBBB-8155-1G
4-(2'-Methoxybenzyloxy)phenylboronsäure
4-(2'-Methoxybenzyloxy)phenylboronsäure
Formel: C₁₄H₁₅BO₄
MDL: MFCD08276786
CAS-Nummer: 871125-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR