3-(4'-Methoxybenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-((4-Methoxybenzyl)oxy)phenyl)boronic acid

BB-8185-1G
COBBBB-8185-1GEA 91.1 EUR
COBBBB-8185-1G
3-(4'-Methoxybenzyloxy)phenylboronsäure
3-(4'-Methoxybenzyloxy)phenylboronsäure
Formel: C₁₄H₁₅BO₄
Molecular Weight: 258,800 g/mol
MDL: MFCD09038416
CAS-Nummer: 1072951-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR