3,5-Diformyl-2-propoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8188-1G BB-8188-250M
COBBBB-8188-1GEA 412 EUR
COBBBB-8188-1G COBBBB-8188-250M
3,5-Diformyl-2-propoxyphenylboronsäure
3,5-Diformyl-2-propoxyphenylboronsäure
Formel: C₁₁H₁₃BO₅
MDL: MFCD09750469
CAS-Nummer: 1072951-92-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR