2-Ethoxy-4-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8190-1G
COBBBB-8190-1GEA 50.8 EUR
COBBBB-8190-1G
2-Ethoxy-4-fluorphenylboronsäure
2-Ethoxy-4-fluorphenylboronsäure
Formel: C₈H₁₀BFO₃
Molecular Weight: 183,97 g/mol
MDL: MFCD03095001
CAS-Nummer: 480438-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR