3-Chlor-4-isopropoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8192-1G
COBBBB-8192-1GEA 74.2 EUR
COBBBB-8192-1G
3-Chlor-4-isopropoxyphenylboronsäure
3-Chlor-4-isopropoxyphenylboronsäure
Formel: C₉H₁₂BClO₃
Molecular Weight: 214.4500 g/mol
MDL: MFCD04974065
CAS-Nummer: 480438-56-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR