2-Fluor-5-iodphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8194-1G
COBBBB-8194-1GEA 0 EUR
COBBBB-8194-1G
2-Fluor-5-iodphenylboronsäure
2-Fluor-5-iodphenylboronsäure
Formel: C₆H₅BFIO₂
MDL: MFCD05664304
CAS-Nummer: 866683-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR