2-Fluor-3-iodphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-8195-1G
COBBBB-8195-1GEA
0
EUR
COBBBB-8195-1G
2-Fluor-3-iodphenylboronsäure
2-Fluor-3-iodphenylboronsäure
Formel:
C₆H₅BFIO₂ |
MDL:
MFCD05664305 CAS-Nummer: 1016231-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...