3-(2'-Methoxybenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8237-1G
COBBBB-8237-1GEA 108 EUR
COBBBB-8237-1G
3-(2'-Methoxybenzyloxy)phenylboronsäure
3-(2'-Methoxybenzyloxy)phenylboronsäure
Formel: C₁₄H₁₅BO₄
MDL: MFCD08276833
CAS-Nummer: 1072952-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR