4-(4'-Methoxybenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-((4-Methoxybenzyl)oxy)phenyl)boronic acid

BB-8240-1G
COBBBB-8240-1GEA 81.3 EUR
COBBBB-8240-1G
4-(4'-Methoxybenzyloxy)phenylboronsäure
4-(4'-Methoxybenzyloxy)phenylboronsäure
Formel: C₁₄H₁₅BO₄
Molecular Weight: 258,0800 g/mol
MDL: MFCD09038417
CAS-Nummer: 156635-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR