4-(Trimethylsilyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(Trimethylsilyl)phenyl)boronic acid

BB-8307-1G
COBBBB-8307-1GEA 63.9 EUR
COBBBB-8307-1G
4-(Trimethylsilyl)phenylboronsäure
4-(Trimethylsilyl)phenylboronsäure
Formel: C₉H₁₅BO₂Si
Molecular Weight: 194,1100 g/mol
MDL: MFCD00093348
CAS-Nummer: 17865-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR