1-Octylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Octylboronic acid, 1-n-Octaneboronic acid, n-Octylboronic acid

BB-8312-5G BB-8312-1G
COBBBB-8312-5GEA 181 EUR
COBBBB-8312-5G COBBBB-8312-1G
1-Octylboronsäure
1-Octylboronsäure
Formel: C₈H₁₉BO₂
Molecular Weight: 158.05 g/mol
MDL: MFCD01074560
CAS-Nummer: 28741-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR