3-Vinylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Vinylbenzolboronsäure, (3-Vinylphenyl)metaborsäure, Dihydroxy(3-vinylphenyl)boran

BB-8320-5G BB-8320-1G
COBBBB-8320-5GEA 273 EUR
COBBBB-8320-5G COBBBB-8320-1G
3-Vinylphenylboronsäure
3-Vinylphenylboronsäure
Formel: C₈H₉BO₂
Molecular Weight: 147.97 g/mol
MDL: MFCD01074679
CAS-Nummer: 15016-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR