2-Formyl-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8334-1G BB-8334-250M
COBBBB-8334-1GEA 142 EUR
COBBBB-8334-1G COBBBB-8334-250M
2-Formyl-5-methylphenylboronsäure
2-Formyl-5-methylphenylboronsäure
Formel: C₈H₉BO₃
Molecular Weight: 163.9663 g/mol
MDL: MFCD01319009
CAS-Nummer: 40138-17-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR