2,3,5-Trichlorphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 2,3,5-Trichlorobenzeneboronic acid, Dihydroxy(2,3,5-trichlorophenyl)borane, (2,3,5-Trichlorophenyl)metaboric acid

BB-8340-1G
COBBBB-8340-1GEA 99.4 EUR
COBBBB-8340-1G
2,3,5-Trichlorphenylboronsäure
2,3,5-Trichlorphenylboronsäure
Formel: C₆H₄BCl₃O₂
Molecular Weight: 225,26 g/mol
MDL: MFCD01630856
CAS-Nummer: 212779-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR