2,4,5-Trifluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8342-5G BB-8342-1G
COBBBB-8342-5GEA 258 EUR
COBBBB-8342-5G COBBBB-8342-1G
2,4,5-Trifluorphenylboronsäure
2,4,5-Trifluorphenylboronsäure
Formel: C₆H₄BF₃O₂
Molecular Weight: 175.9 g/mol
MDL: MFCD01863165
CAS-Nummer: 247564-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR