2,4,5-Trifluorphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-8342-5G
BB-8342-1G
COBBBB-8342-5GEA
258
EUR
COBBBB-8342-5G
COBBBB-8342-1G
2,4,5-Trifluorphenylboronsäure
2,4,5-Trifluorphenylboronsäure
Formel:
C₆H₄BF₃O₂ Molecular Weight: 175.9 g/mol |
MDL:
MFCD01863165 CAS-Nummer: 247564-72-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...