2-(Hydroxymethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Hydroxymethyl)phenyl)boronic acid, 2-Hydroxymethylphenylboronic acid, 2-(Hydroxymethyl)benzeneboronic acid

BB-8356-5G BB-8356-1G BB-8356-25G
COBBBB-8356-5GEA 0 EUR
COBBBB-8356-5G COBBBB-8356-1G COBBBB-8356-25G
2-(Hydroxymethyl)phenylboronsäure
2-(Hydroxymethyl)phenylboronsäure
Formel: C₇H₉BO₃
MDL: MFCD02258966
CAS-Nummer: 87199-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR