2-Formyl-5-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Methoxy-2-formylphenylboronic acid

BB-8361-1G
COBBBB-8361-1GEA 127 EUR
COBBBB-8361-1G
2-Formyl-5-methoxyphenylboronsäure
2-Formyl-5-methoxyphenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179,9657 g/mol
MDL: MFCD03001335
CAS-Nummer: 40138-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR