3-(Trimethylsilyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-(Trimethylsilyl)phenyl)boronic acid
BB-8371-1G
COBBBB-8371-1GEA
95.9
EUR
COBBBB-8371-1G
3-(Trimethylsilyl)phenylboronsäure
3-(Trimethylsilyl)phenylboronsäure
Formel:
C₉H₁₅BO₂Si Molecular Weight: 194,1100 g/mol |
MDL:
MFCD03093884 CAS-Nummer: 177171-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...