2,6-Dichlor-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2,6-Dichloropyridin-3-ylboronic acid, 2,6-Dichloropyridine-3-boronic acid
BB-8377-1G
COBBBB-8377-1GEA
38.5
EUR
COBBBB-8377-1G
2,6-Dichlor-3-pyridinboronsäure
2,6-Dichloro-3-pyridineboronic acid
Formel:
C₅H₄BCl₂NO₂ Molecular Weight: 191,81 g/mol |
MDL:
MFCD03094989 CAS-Nummer: 148493-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...