2-(1-Piperidinyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-(Piperidin-1-yl)phenyl)boronic acid, 2-(Piperidino)phenylboronic acid
BB-8382-1G
COBBBB-8382-1GEA
0
EUR
COBBBB-8382-1G
2-(1-Piperidinyl)phenylboronsäure
2-(1-Piperidinyl)phenylboronsäure
Formel:
C₁₁H₁₆BNO₂ |
MDL:
MFCD03095111 CAS-Nummer: 1050505-85-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...