3-(Ethylsulfonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8397-1G
COBBBB-8397-1GEA 0 EUR
COBBBB-8397-1G
3-(Ethylsulfonyl)phenylboronsäure
3-(Ethylsulfonyl)phenylboronsäure
Formel: C₈H₁₁BO₄S
MDL: MFCD03095257
CAS-Nummer: 845870-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR