2,4-Dichlor-5-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8399-1G
COBBBB-8399-1GEA 0 EUR
COBBBB-8399-1G
2,4-Dichlor-5-nitrophenylboronsäure
2,4-Dichlor-5-nitrophenylboronsäure
Formel: C₆H₄BCl₂NO₄
MDL: MFCD03095325
CAS-Nummer: 1072952-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR