4-(Piperidin-1-yl)phenylboronsäure Hydrochlorid 95

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Piperidino)phenylboronic acid hydrochloride, (4-(Piperidin-1-yl)phenyl)boronic acid hydrochloride

BB-8409-1G BB-8409-250M
COBBBB-8409-1GEA 864 EUR
COBBBB-8409-1G COBBBB-8409-250M
4-(Piperidin-1-yl)phenylboronsäure Hydrochlorid 95
4-(Piperidin-1-yl)phenylboronsäure Hydrochlorid
Formel: C₁₁H₁₇BClNO₂
Molecular Weight: 205.06 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD03410255
CAS-Nummer: 229009-42-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR