2-Fluor-6-methyl-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-6-methylpyridine-3-boronic acid, 2-Fluoro-6-methylpyridin-3-ylboronic acid

BB-8411-1G
COBBBB-8411-1GEA 303 EUR
COBBBB-8411-1G
2-Fluor-6-methyl-3-pyridinboronsäure
2-Fluoro-6-methyl-3-pyridineboronic acid
Formel: C₆H₇BFNO₂
Molecular Weight: 154,94 g/mol
MDL: MFCD03411560
CAS-Nummer: 906744-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR