2-Chlor-6-methyl-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-8414-1G
COBBBB-8414-1GEA 451 EUR
COBBBB-8414-1G
2-Chlor-6-methyl-3-pyridinboronsäure
2-Chloro-6-methyl-3-pyridineboronic acid
Formel: C₆H₇BClNO₂
MDL: MFCD03411563
CAS-Nummer: 536693-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR