2-Chlor-3-methyl-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-8415-5G BB-8415-1G
COBBBB-8415-5GEA 0 EUR
COBBBB-8415-5G COBBBB-8415-1G
2-Chlor-3-methyl-5-pyridinboronsäure
2-Chlor-3-methyl-5-pyridinboronsäure
Formel: C₆H₇BClNO₂
MDL: MFCD03411564
CAS-Nummer: 1003043-40-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR