3-(Morpholino)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Morpholinophenyl)boronic acid
BB-8418-1G
COBBBB-8418-1GEA
154
EUR
COBBBB-8418-1G
3-(Morpholino)phenylboronsäure
3-(Morpholino)phenylboronsäure
Formel:
C₁₀H₁₄BNO₃ Molecular Weight: 207,0341 g/mol |
MDL:
MFCD03412095 CAS-Nummer: 863377-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...