2-Butoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8423-1G
COBBBB-8423-1GEA 50.9 EUR
COBBBB-8423-1G
2-Butoxyphenylboronsäure
2-Butoxyphenylboronsäure
Formel: C₁₀H₁₅BO₃
Molecular Weight: 194,04 g/mol
MDL: MFCD03427052
CAS-Nummer: 91129-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR