3-Propoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8425-1G
COBBBB-8425-1GEA 37.4 EUR
COBBBB-8425-1G
3-Propoxyphenylboronsäure
3-Propoxyphenylboronsäure
Formel: C₉H₁₃BO₃
Molecular Weight: 180,0100 g/mol
MDL: MFCD03427190
CAS-Nummer: 149557-18-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR