2,3,4,6-Tetrafluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Dihydroxy(2,3,4,6-tetrafluorphenyl)boran, 2,3,4,6-Tetrafluorbenzolboronsäure, (2,3,4,6-Tetrafluorphenyl)metaborsäure

BB-8454-1G
COBBBB-8454-1GEA 204 EUR
COBBBB-8454-1G
2,3,4,6-Tetrafluorphenylboronsäure
2,3,4,6-Tetrafluorphenylboronsäure
Formel: C₆H₃BF₄O₂
Molecular Weight: 193.89 g/mol
MDL: MFCD04039309
CAS-Nummer: 511295-00-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR