2,3,5,6-Tetrafluorphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 2,3,5,6-Tetrafluorbenzolboronsäure, (2,3,5,6-Tetrafluorphenyl)metaborsäure, Dihydroxy(2,3,5,6-tetrafluorphenyl)boran

BB-8455-1G
COBBBB-8455-1GEA 581 EUR
COBBBB-8455-1G
2,3,5,6-Tetrafluorphenylboronsäure
2,3,5,6-Tetrafluorphenylboronsäure
Formel: C₆H₃BF₄O₂
Molecular Weight: 193,89 g/mol
MDL: MFCD04039310
CAS-Nummer: 511295-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR