2,6-Difluor-4-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2,6-Difluoropyridin-4-yl)boronic acid, 2,6-Difluoro-4-pyridylboronic acid
BB-8468-250M
COBBBB-8468-250MEA
288
EUR
COBBBB-8468-250M
2,6-Difluor-4-pyridinboronsäure
2,6-Difluoro-4-pyridineboronic acid
Formel:
C₅H₄BF₂NO₂ Molecular Weight: 158,8986 g/mol |
MDL:
MFCD04114289 CAS-Nummer: 401816-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...