2-Fluor-5-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8476-5G BB-8476-25G BB-8476-1G
COBBBB-8476-5GEA 50.9 EUR
COBBBB-8476-5G COBBBB-8476-25G COBBBB-8476-1G
2-Fluor-5-methoxyphenylboronsäure
2-Fluor-5-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
Molecular Weight: 169.9460 g/mol
MDL: MFCD04974115
CAS-Nummer: 406482-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR