2-Chlor-3-thiophenboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Chlorothiophen-3-yl)boronic acid

BB-8478-1G
COBBBB-8478-1GEA 182 EUR
COBBBB-8478-1G
2-Chlor-3-thiophenboronsäure
2-Chloro-3-thiopheneboronic acid
Formel: C₄H₄BClO₂S
Molecular Weight: 162,4 g/mol
MDL: MFCD05662391
CAS-Nummer: 177734-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR