5-Chlor-2-fluor-3-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-Chloro-2-fluoro-3-methylphenyl)boronic acid, 5-Chloro-2-fluoro-3-methylboronic acid

BB-8482-1G
COBBBB-8482-1GEA 121 EUR
COBBBB-8482-1G
5-Chlor-2-fluor-3-methylphenylboronsäure
5-Chlor-2-fluor-3-methylphenylboronsäure
Formel: C₇H₇BClFO₂
Molecular Weight: 188,391 g/mol
MDL: MFCD05664227
CAS-Nummer: 352535-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR