N,N,2-Trimethylanilin-4-boronsäure

Lieferant: COMBI-BLOCKS

BB-8497-1G
COBBBB-8497-1GEA 0 EUR
COBBBB-8497-1G
N,N,2-Trimethylanilin-4-boronsäure
N,N,2-Trimethylanilin-4-boronsäure
Formel: C₉H₁₄BNO₂
MDL: MFCD06657910
CAS-Nummer: 919496-59-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR