2-Methoxy-3-pyridinboronsäure Hydrat

Lieferant: COMBI-BLOCKS

BB-8498-5G BB-8498-25G
COBBBB-8498-5GEA 0 EUR
COBBBB-8498-5G COBBBB-8498-25G
2-Methoxy-3-pyridinboronsäure Hydrat
2-Methoxy-3-pyridinboronsäure Hydrat
Formel: C₆H₈BNO₃·H₂O
MDL: MFCD06659055
CAS-Nummer: 1256355-25-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR