2-Methyl-3-pyridinboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

BB-8500-1G
COBBBB-8500-1GEA 0 EUR
COBBBB-8500-1G
2-Methyl-3-pyridinboronsäure Hydrochlorid
2-Methyl-3-pyridinboronsäure Hydrochlorid
Formel: C₆H₉BClNO₂
MDL: MFCD06659489
CAS-Nummer: 1072952-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR