3-Fluor-5-methyl-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-8518-1G
COBBBB-8518-1GEA 742 EUR
COBBBB-8518-1G
3-Fluor-5-methyl-4-pyridinboronsäure
3-Fluoro-5-methyl-4-pyridineboronic acid
Formel: C₆H₇BFNO₂
MDL: MFCD07368843
CAS-Nummer: 1072952-44-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR